Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Histone H2B type 1B Antibody, Novus Biologicals™
SDP

Catalog No. NB387857 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB387857 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB387857 Supplier Novus Biologicals Supplier No. NBP323376
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Histone H2B type 1B Polyclonal antibody specifically detects Histone H2B type 1B in Human, Mouse, Rat, Xenopus samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen Histone H2B type 1B
Applications Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:5000, Immunocytochemistry/ Immunofluorescence 1:200
Formulation PBS, 1 mg/mL BSA and 50% Glycerol
Gene Alias H2B histone family, member F, H2B/fHistone H2B.f, H2BFFH2B.1, histone 1, H2bb, histone cluster 1, H2bb, histone H2B type 1-B, Histone H2B.1, MGC119804
Host Species Rabbit
Immunogen Histone H2B synthetic peptide (coupled to carrier protein) corresponds to amino acids in the C-terminus of human histone H2B. This sequence is well conserved in rat, mouse, fish, and drosophila histone H2B, and has low homology to other histone family members. [UniProt# P33778]
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Chromatin Research, Epigenetics
Primary or Secondary Primary
Gene ID (Entrez) 3018
Target Species Human, Mouse, Rat, Xenopus species
Content And Storage Store at -20°C.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.