Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Histone H2B type 2E Antibody, Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB086237 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB086237 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB086237 Supplier Novus Biologicals Supplier No. NBP299782AF488
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Histone H2B type 2E Polyclonal specifically detects Histone H2B type 2E in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Histone H2B type 2E
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 488
Formulation 50 mM Sodium Borate
Gene Alias GL105, H2B, H2B Histone Family, Member Q, H2B.1, Histone 2, H2be, Histone Cluster 2 H2B Family Member E, Histone Cluster 2, H2be, Histone H2B Type 2-E, Histone H2B.Q, Histone H2B-GL105
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Histone H2B type 2E (Q16778; Met1-Lys126).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8349
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.