Learn More
Description
Specifically detects Histone H2B in Human, Mouse samples, and it is validated for Western Blotting
Specifications
Specifications
| Antigen | HIST2H2BE |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500-1:3000 |
| Formulation | PBS (pH 7.0) and 20% Glycerol with 0.025% Proclin 300 |
| Gene Symbols | H2BC21 |
| Host Species | Rabbit |
| Immunogen | Full length human Histone H2B Recombinant protein. |
| Molecular Weight of Antigen | 14 kDa |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
