Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Histone H2B type 2E Antibody (SR1176), Novus Biologicals™
SDP

Catalog No. NB258844 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB258844 100 μg
NB258847 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB258844 Supplier Novus Biologicals Supplier No. NBP322445100UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Histone H2B type 2E Monoclonal antibody specifically detects Histone H2B type 2E in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry

Specifications

Antigen Histone H2B type 2E
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone SR1176
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200
Formulation PBS, pH 7.4, 150mM NaCl, 50% glycerol.
Gene Alias GL105, H2B, H2B Histone Family, Member Q, H2B.1, Histone 2, H2be, Histone Cluster 2 H2B Family Member E, Histone Cluster 2, H2be, Histone H2B Type 2-E, Histone H2B.Q, Histone H2B-GL105
Host Species Rabbit
Immunogen A synthesized peptide derived from human Histone H2B type 2E (Uniprot #: Q16778)
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Chromatin Research, Epigenetics
Primary or Secondary Primary
Gene ID (Entrez) 8349
Target Species Human, Mouse, Rat
Content And Storage Store at -20° C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.