Learn More
Histone H2B type 2E Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | Histone H2B type 2E |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol, pH7.3. |
| Gene Alias | GL105, H2B, H2B Histone Family, Member Q, H2B.1, Histone 2, H2be, Histone Cluster 2 H2B Family Member E, Histone Cluster 2, H2be, Histone H2B Type 2-E, Histone H2B.Q, Histone H2B-GL105 |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-126 of human Histone H2B type 2E (NP_003519.1). |
| Purification Method | Affinity purified |
| Quantity | 20 μL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.