Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Histone H2B, ubiquitylated Lys123 Antibody (1B3F12/A9) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody has been used in 1 publication
Supplier: Novus Biologicals NBP254770
Description
Histone H2B Monoclonal specifically detects Histone H2B in Human, S. pombe, Yeast samples. It is validated for Western Blot, Chromatin Immunoprecipitation, ELISA, Immunocytochemistry/Immunofluorescence, Chromatin Immunoprecipitation (ChIP).Specifications
Histone H2B | |
Monoclonal | |
1.0 mg/mL | |
Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence, Chromatin Immunoprecipitation (ChIP) 1:10-1:500 | |
FLJ35099, FLJ56780, FLJ56787, histone 2, H2bf, histone cluster 2, H2bf, histone H2B type 2-F, MGC131639 | |
Mouse | |
23 kDa | |
0.1 mL | |
Chromatin Research, Epigenetics, Histones and Modified Histones | |
440689 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG1 |
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence, ChIP Assay | |
1B3F12/A9 | |
Unconjugated | |
PBS with 0.02% Sodium Azide | |
HIST2H2BF | |
Ub-polypeptide antigen based on amino acids 115 - 130 of yeast Histone H2B | |
Protein A purified | |
RUO | |
Primary | |
Human, S. pombe, Yeast | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction