Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Histone H2B, ubiquitylated Lys123 Antibody (1B3F12/A9) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody has been used in 1 publication
$196.10 - $499.50
Specifications
Antigen | Histone H2B |
---|---|
Clone | 1B3F12/A9 |
Concentration | 1.0 mg/mL |
Dilution | Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence, Chromatin Immunoprecipitation (ChIP) 1:10-1:500 |
Applications | Western Blot, ELISA, Immunocytochemistry, Immunofluorescence, ChIP Assay |
Description
Histone H2B Monoclonal specifically detects Histone H2B in Human, S. pombe, Yeast samples. It is validated for Western Blot, Chromatin Immunoprecipitation, ELISA, Immunocytochemistry/Immunofluorescence, Chromatin Immunoprecipitation (ChIP).Specifications
Histone H2B | |
1.0 mg/mL | |
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence, ChIP Assay | |
Unconjugated | |
Mouse | |
Chromatin Research, Epigenetics, Histones and Modified Histones | |
PBS with 0.02% Sodium Azide | |
440689 | |
Ub-polypeptide antigen based on amino acids 115 - 130 of yeast Histone H2B | |
Primary | |
23 kDa |
1B3F12/A9 | |
Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence, Chromatin Immunoprecipitation (ChIP) 1:10-1:500 | |
Monoclonal | |
Purified | |
RUO | |
Human, S. pombe, Yeast | |
FLJ35099, FLJ56780, FLJ56787, histone 2, H2bf, histone cluster 2, H2bf, histone H2B type 2-F, MGC131639 | |
HIST2H2BF | |
IgG1 | |
Protein A purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title