Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Histone H2B, ubiquitylated Lys123 Antibody (1B3F12/A9), mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB309112 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB309112 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB309112 Supplier Novus Biologicals Supplier No. NBP254770MFV500
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Histone H2B Monoclonal antibody specifically detects Histone H2B in Human, S. pombe, Yeast samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, ChIP assay (ChIP)

Specifications

Antigen Histone H2B
Applications Western Blot, ELISA, Immunocytochemistry, ChIP Assay
Classification Monoclonal
Clone 1B3F12/A9
Conjugate mFluor Violet 500 SE
Dilution Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Chromatin Immunoprecipitation (ChIP)
Formulation 50mM Sodium Borate
Gene Alias FLJ35099, FLJ56780, FLJ56787, histone 2, H2bf, histone cluster 2, H2bf, histone H2B type 2-F, MGC131639
Host Species Mouse
Immunogen Ub-polypeptide antigen based on amino acids 115 - 130 of yeast Histone H2B
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Chromatin Research, Epigenetics
Primary or Secondary Primary
Gene ID (Entrez) 440689
Target Species Human, S. pombe, Yeast
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.