Learn More
Histone H2B, ubiquitylated Lys123 Antibody (C.15200232), Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | Histone H2B (ubiquitylated Lys123) |
| Applications | Western Blot, ChIP Assay |
| Classification | Monoclonal |
| Clone | C.15200232 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500, Chromatin Immunoprecipitation (ChIP) 2 ug/IP |
| Formulation | PBS with 0.02% Sodium Azide |
| Gene Alias | FLJ35099, FLJ56780, FLJ56787, histone 2, H2bf, histone cluster 2, H2bf, histone H2B type 2-F, MGC131639 |
| Gene Symbols | HIST2H2BF |
| Host Species | Mouse |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.