Learn More
Histone H2BK12ac Polyclonal Antibody, Active Motif™

Description
Specifications
Specifications
| Antigen | Histone H2BK12ac |
| Applications | ChIP Assay, Dot Blot, Immunocytochemistry, Immunofluorescence, Western Blot |
| Classification | Polyclonal |
| Concentration | Conc. Not Determined |
| Conjugate | Unconjugated |
| Formulation | whole serum with 30% glycerol and 0.035% sodium azide; pH |
| Gene | H2BC12 |
| Gene Accession No. | O60814, P23527, P62807, Q16778, Q5QNW6, Q6DN03, Q6DRA6, Q99877 |
| Gene Alias | 2610022J01Rik; Acetyl-Histone H2; Acetyl-Histone H2B K12; Acetyl-Histone H2B Lys12; AV127319; dJ193B12.2; dJ221C16.3; dJ221C16.8; GL105; H2B; H2b 613; H2B GL105; H2B histone family, member A; H2B histone family, member D; H2B histone family, member K; H2B histone family, member L; H2B histone family, member N; H2B histone family, member Q; H2B histone family, member S; H2B histone family, member T; H2B K; H2B.1; H2B.1A; H2B.2; H2B/a; H2B/d; H2B/g; H2B/h; H2B/I; H2B/k; H2B/l; H2B/n; H2B/o; H2B/q; H2B/s; H2B/t; H2b-613; H2BC10; H2bc4; H2bc6; H2BC7; H2bc8; H2BFA; H2BFAiii; H2BFD; H2BFG; H2BFH; H2BFK; H2BFL; H2BFN; H2BFO; H2BFQ; H2bfs; H2BFT; H2BGL105; H2BK; H2BK12ac; H2BK15ac; H2BK5ac; H2BLys12ac; H2BQ; H2BS14; H2BS14p; HIRA-interacting protein 1; HIRIP1; Hist1h2ba; HIST1H2BC; HIST1H2BE; HIST1H2BF; Hist1h2bg; HIST1H2BI; Hist1h2bk; HIST1H2BN; HIST1H2BO; HIST1H3I; HIST2H2BC; HIST2H2BD; Hist2h2be; HIST2H2BF; histone 1, H2ba; histone 1, H2bc; histone 1, H2bg; histone 1, H2bi; histone 1, H2bk; histone 1, H2bn; histone 1, H2bo; histone 2, H2be; Histone 2B; histone cluster 1, H2ba; histone cluster 1, H2bc; histone cluster 1, H2bg; histone cluster 1, H2bi; histone cluster 1, H2bk; histone cluster 1, H2bn; histone cluster 1, H2bo; histone cluster 2, H2bc (pseudogene); histone cluster 2, H2bd (pseudogene); histone cluster 2, H2be; histone cluster 2, H2bf; histone family member; histone H2B; Histone H2B type 1-A; Histone H2B type 1-C/E/F/G/I; Histone H2B type 1-C/E/G; Histone H2B type 1-K; Histone H2B type 1-N; Histone H2B type 1-O; Histone H2B type 2-E; Histone H2B type 2-F; Histone H2B, testis; Histone H2B.1 A; Histone H2B.2; Histone H2B.a; Histone H2B.d; Histone H2B.g; Histone H2B.h; Histone H2B.I; Histone H2B.k; Histone H2B.l; Histone H2B.n; Histone H2B.q; Histone H2B-GL105; MGC125414; MGC125415; MGC125416; MGC9388; R74621; RP1-193B12.10; T25626; testis-specific histone 2b; Testis-specific histone H2B; Th2b |
| Gene Symbols | H2BC10, H2BC12, H2BC15, H2BC17, H2BC18, H2BC19P, H2BC20P, H2BC21, H2BC4, H2BC8 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.