Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Histone H2BK15ac Polyclonal Antibody, Active Motif™

Rabbit Polyclonal Antibody
$156.65 - $620.58
Specifications
Antigen | Histone H2BK15ac |
---|---|
Concentration | Lot-Specific |
Applications | Dot Blot, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-199-3329
![]() ![]() |
Active Motif
61322 |
10 μL |
Each for $156.65
|
|
|||||
50-199-3328
![]() ![]() |
Active Motif
61321 |
100 μL |
Each for $620.58
|
|
|||||
Description
Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes. Nucleosomes consist of approximately 146 bp of DNA wrapped around a histone octamer composed of pairs of each of the four core histones (H2A, H2B, H3, and H4). The nucleosomes wrap further and compact DNA into chromotin, limiting DNA accessibility to the cellular machineries which require DNA as a template. Histones play a central role in transciption regulation, DNA repair, DNA replication, and chromosomal stability. H2B has a broad antibacterial activity.Specifications
Histone H2BK15ac | |
Dot Blot, Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
O60814, P23527, P62807, Q16778, Q5QNW6, Q6DN03, Q6DRA6, Q99877 | |
337873, 337874, 440689, 8339, 8341, 8346, 8347, 8348, 8349, 85236 | |
This antibody was raised against a peptide including acetyl-lysine 15 of human histone H2B. | |
Primary | |
-20°C, Avoid Freeze/Thaw Cycles | |
H2BC12 |
Lot-Specific | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 30% glycerol and 0.035% sodium azide; pH 7.5 | |
2610022J01Rik; Acetyl-Histone H2; Acetyl-Histone H2B K15; Acetyl-Histone H2B Lys15; AV127319; dJ193B12.2; dJ221C16.3; dJ221C16.8; GL105; H2B; H2b 613; H2B GL105; H2B histone family, member A; H2B histone family, member D; H2B histone family, member K; H2B histone family, member L; H2B histone family, member N; H2B histone family, member Q; H2B histone family, member S; H2B histone family, member T; H2B K; H2B.1; H2B.1A; H2B.2; H2B/a; H2B/d; H2B/g; H2B/h; H2B/I; H2B/k; H2B/l; H2B/n; H2B/o; H2B/q; H2B/s; H2B/t; H2b-613; H2BC10; H2BC4; H2BC6; H2BC7; H2bc8; H2BFA; H2BFAiii; H2BFD; H2BFG; H2BFH; H2BFK; H2BFL; H2BFN; H2BFO; H2BFQ; H2bfs; H2BFT; H2BGL105; H2BK; H2BK12ac; H2BK15ac; H2BK5ac; H2BLys15ac; H2BQ; H2BS14; H2BS14p; HIRA-interacting protein 1; HIRIP1; Hist1h2ba; Hist1h2bc; Hist1h2be; HIST1H2BF; HIST1H2BG; HIST1H2BI; Hist1h2bk; HIST1H2BN; HIST1H2BO; HIST1H3I; HIST2H2BC; HIST2H2BD; HIST2H2BE; HIST2H2BF; histone 1, H2ba; histone 1, H2bc; histone 1, H2bg; histone 1, H2bi; histone 1, H2bk; histone 1, H2bn; histone 1, H2bo; histone 2, H2be; Histone 2B; histone cluster 1, H2ba; histone cluster 1, H2bc; histone cluster 1, H2bg; histone cluster 1, H2bi; histone cluster 1, H2bk; histone cluster 1, H2bn; histone cluster 1, H2bo; histone cluster 2, H2bc (pseudogene); histone cluster 2, H2bd (pseudogene); histone cluster 2, H2be; histone cluster 2, H2bf; histone family member; histone H2B; Histone H2B type 1-A; Histone H2B type 1-C/E/F/G/I; Histone H2B type 1-C/E/G; Histone H2B type 1-K; Histone H2B type 1-N; Histone H2B type 1-O; Histone H2B type 2-E; Histone H2B type 2-F; Histone H2B, testis; Histone H2B.1 A; Histone H2B.2; Histone H2B.a; Histone H2B.d; Histone H2B.g; Histone H2B.h; Histone H2B.I; Histone H2B.k; Histone H2B.l; Histone H2B.n; Histone H2B.q; Histone H2B-GL105; MGC125414; MGC125415; MGC125416; MGC9388; R74621; RP1-193B12.10; T25626; testis-specific histone 2b; Testis-specific histone H2B; Th2b | |
H2BC10, H2BC12, H2BC15, H2BC17, H2BC18, H2BC19P, H2BC20P, H2BC21, H2BC4, H2BC8 | |
IgG | |
Antigen affinity chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title