Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Histone H3.3 Antibody, Novus Biologicals™
SDP

Catalog No. NBP224697SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP224697SS 0.025 mg
NBP224697 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP224697SS Supplier Novus Biologicals Supplier No. NBP224697SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 1 publication

Histone H3.3 Polyclonal specifically detects Histone H3.3 in Human, Mouse, Bovine, Chicken, Drosophila, Equine, Opossum, Zebrafish samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Histone H3.3
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1.0 mg/ml
Conjugate Unconjugated
Dilution Western Blot 0.25-0.5 ug/ml, Immunohistochemistry-Paraffin 5-10 ug/ml
Formulation PBS with 0.05% Sodium Azide
Gene Alias H3.3A, H3F3, H3F3A, Histone H3.3, Histone H3.3A, Histone H3.3B, MGC87782, MGC87783
Gene Symbols H3F3A
Host Species Rabbit
Immunogen A portion of amino acids 100-150 of human Histone H3.3 was used as the immunogen for the antibody.
Purification Method Protein A purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Chromatin Modifiers, Chromatin Research, Epigenetics, Histones and Modified Histones
Primary or Secondary Primary
Gene ID (Entrez) 3020
Test Specificity The Histone H3.3 amino acid sequence used as immunogen for the antibody is 100% homologous in human (Histone H3.3A, B, and C), Drosophila, orangutan, horse, oppossum, mouse, rat, cow, chicken, Xenopus, salmon and zebrafish.
Target Species Human, Mouse, Bovine, Chicken, Drosophila, Equine, Opossum
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.