Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Histone H3.3A Antibody, DyLight 405, Novus Biologicals™
SDP

Catalog No. NB055329 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB055329 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB055329 Supplier Novus Biologicals Supplier No. NBP298769V
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Histone H3.3A Polyclonal specifically detects Histone H3.3A in Human samples. It is validated for Western Blot.

Specifications

Antigen Histone H3.3A
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 405
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias H3 histone, family 3A, H3.3AH3F3H3F3B, H3.3B, histone H3.3, MGC87782, MGC87783
Gene Symbols H3-3A
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of human Histone H3, Family 3A.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3020
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.