Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Histone H3, ac Lys14 Antibody (RM322) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB301043 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB301043 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB301043 Supplier Novus Biologicals Supplier No. NBP326006
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Histone H3 Monoclonal antibody specifically detects Histone H3 in Human, Vertebrate samples. It is validated for Western Blot, Multiplex Immunoassay

Specifications

Antigen Histone H3
Applications Western Blot, Multiplex
Classification Monoclonal
Clone RM322
Conjugate Unconjugated
Dilution Western Blot : 0.1 - 1 ug/mL, Multiplex Immunoassay : 0.01 - 1 ug/mL
Formulation PBS
Gene Alias H3, H3 histone, family 3A, H3.3A, H3.3B, H3F2, H3F3, H3F3B, H3FM, HIST2H3C, histone H3.3, MGC87782, MGC87783
Host Species Rabbit
Immunogen A dimethyl-peptide corresponding to Histone H3 [Lys9] and an acetyl-peptide corresponding to Histone H3.3B [Lys14] (Uniprot # P84243)
Purification Method Protein A purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Chromatin Modifiers, Chromatin Research, Epigenetics, Histones and Modified Histones, Loading Controls
Primary or Secondary Primary
Gene ID (Entrez) 126961
Target Species Human, Vertebrates
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.