Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Histone H3, ac Lys9 Antibody, Novus Biologicals™
SDP

Catalog No. NBP259180 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP259180 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP259180 Supplier Novus Biologicals Supplier No. NBP259180
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Histone H3 Polyclonal specifically detects Histone H3 in Human, Mouse, P. falciparum, Plant, Zebrafish samples. It is validated for Western Blot, Chromatin Immunoprecipitation, ELISA, Dot Blot, Immunofluorescence, Immunocytochemistry, Chromatin Immunoprecipitation Sequencing.

Specifications

Antigen Histone H3 (ac Lys9)
Applications Western Blot, ELISA, Dot Blot, Immunofluorescence, ChIP Assay
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000, ELISA 1:1000, Dot Blot 1:20000, Immunofluorescence 1:500, Chromatin Immunoprecipitation (ChIP) 1-2 ug/IP, Immunocytochemistry 1:500, Chromatin Immunoprecipitation Sequencing
Formulation PBS with 0.05% Sodium Azide and 0.05% ProClin 300
Gene Alias H3, H3 histone, family 3A, H3.3A, H3.3B, H3F2, H3F3, H3F3B, H3FM, H3K9ac, HIST2H3C, histone H3.3, MGC87782, MGC87783
Gene Symbols H3C14
Host Species Rabbit
Immunogen The exact sequence of the immunogen to this Histone H3 [ac Lys9] antibody is proprietary.
Molecular Weight of Antigen 15 kDa
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Chromatin Modifiers, Chromatin Research, Epigenetics, Histones and Modified Histones, Loading Controls
Primary or Secondary Primary
Gene ID (Entrez) 126961
Target Species Human, Mouse, P. falciparum, Plant, Zebrafish
Content And Storage Store at -20C short term. Aliquot and store at -80C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.