Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Histone H3 Antibody, Novus Biologicals™
SDP

Catalog No. NB500267SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB500267SS 0.025 mL
NB500267 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB500267SS Supplier Novus Biologicals Supplier No. NB500267SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 3 publications

Histone H3 Polyclonal specifically detects Histone H3 in Human, Mouse samples. It is validated for Western Blot.

Specifications

Antigen Histone H3
Applications Western Blot, ChIP Assay
Classification Polyclonal
Concentration 18.0mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:1000, Chromatin Immunoprecipitation reported in scientific literature (PMID 35031618), Chromatin Immunoprecipitation (ChIP)
Formulation PBS with 0.02% Sodium Azide
Gene Alias H3, H3 histone, family 3A, H3.3A, H3.3B, H3F2, H3F3, H3F3B, H3FM, HIST2H3C, histone H3.3, MGC87782, MGC87783
Gene Symbols H3C14
Host Species Rabbit
Immunogen This Histone H3 antibody was raised against a synthetic peptide corresponding to a sequence within amino acids 100-135 of human Histone H3.
Molecular Weight of Antigen 15 kDa
Purification Method Ammonium sulfate precipitation
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Chromatin Modifiers, Chromatin Research, Epigenetics, Histones and Modified Histones, Loading Controls
Primary or Secondary Primary
Gene ID (Entrez) 126961
Test Specificity Human and mouse. Predicted to cross-react with all mammals and a wide range of other species due to sequence homology.
Target Species Human, Mouse
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Precipitation
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.