Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Histone H3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 3 publications
$206.50 - $499.50
Specifications
Antigen | Histone H3 |
---|---|
Concentration | 18.0mg/mL |
Dilution | Western Blot 1:500-1:1000, Chromatin Immunoprecipitation reported in scientific literature (PMID 35031618), Chromatin Immunoprecipitation (ChIP) |
Applications | Western Blot, ChIP Assay |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB500267SS
![]() |
Novus Biologicals
NB500267SS |
0.025 mL |
Each for $206.50
|
|
|||||
NB500267
![]() |
Novus Biologicals
NB500267 |
0.1 mL |
Each for $499.50
|
|
|||||
Description
Histone H3 Polyclonal specifically detects Histone H3 in Human, Mouse samples. It is validated for Western Blot.Specifications
Histone H3 | |
Western Blot 1:500-1:1000, Chromatin Immunoprecipitation reported in scientific literature (PMID 35031618), Chromatin Immunoprecipitation (ChIP) | |
Polyclonal | |
Precipitation | |
RUO | |
Human, Mouse | |
H3, H3 histone, family 3A, H3.3A, H3.3B, H3F2, H3F3, H3F3B, H3FM, HIST2H3C, histone H3.3, MGC87782, MGC87783 | |
H3C14 | |
IgG | |
Ammonium sulfate precipitation | |
Human and mouse. Predicted to cross-react with all mammals and a wide range of other species due to sequence homology. |
18.0mg/mL | |
Western Blot, ChIP Assay | |
Unconjugated | |
Rabbit | |
Chromatin Modifiers, Chromatin Research, Epigenetics, Histones and Modified Histones, Loading Controls | |
PBS with 0.02% Sodium Azide | |
126961 | |
This Histone H3 antibody was raised against a synthetic peptide corresponding to a sequence within amino acids 100-135 of human Histone H3. | |
Primary | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
15 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title