Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Histone H3 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB500171028 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB500171028 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB500171028 Supplier Novus Biologicals Supplier No. NB5001710.025ML
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 12 publications

Histone H3 Polyclonal specifically detects Histone H3 in Human, Mouse, Rat, Insect, Xenopus, Yeast samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunoblotting, Single Cell Western.

Specifications

Antigen Histone H3
Applications ChIP Assay, Immunocytochemistry, Immunofluorescence, Immunohistochemistry
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:4000, Chromatin Immunoprecipitation, Immunohistochemistry 1:100-1:300, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:100-1:300
Formulation PBS (pH 7.4) with 0.01% Sodium Azide
Gene Accession No. P68431
Gene Alias H3 histone, family 3A, H3.3AH3F3H3F3B, H3.3B, histone H3.3, MGC87782, MGC87783
Gene Symbols HIST1H3F
Host Species Rabbit
Immunogen A synthetic peptide made to an N-terminal portion of the human Histone H3 protein (between residues 100-150) [UniProt P68431]
Purification Method Affinity Purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Chromatin Research, Epigenetics, Histones and Modified Histones, Loading Controls
Primary or Secondary Primary
Gene ID (Entrez) 126961
Target Species Human, Mouse, Rat, Xenopus species, Yeast
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.