Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Histone H3 Antibody (C.15200187), Novus Biologicals™
SDP

Catalog No. NBP259304 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP259304 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP259304 Supplier Novus Biologicals Supplier No. NBP259304

Mouse Monoclonal Antibody

Histone H3 Monoclonal specifically detects Histone H3 in Human samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Histone H3
Applications ChIP Assay, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone C.15200187
Conjugate Unconjugated
Dilution Western Blot 1-2 ug, Chromatin Immunoprecipitation 1:1000, Immunocytochemistry/Immunofluorescence
Formulation PBS with No Preservative
Gene Alias H3, H3 histone, family 3A, H3.3A, H3.3B, H3F2, H3F3, H3F3B, H3FM, HIST2H3C, histone H3.3, MGC87782, MGC87783
Gene Symbols H3C14
Host Species Mouse
Immunogen This Histone H3 antibody (C.15200187) was raised against monoclonal antibody raised in mouse against a region of histone H3 containing the unmodified lysine 9 (H3K9un), using a KLH-conjugated synthetic peptide.
Molecular Weight of Antigen 15 kDa
Purification Method Protein A purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Chromatin Modifiers, Chromatin Research, Epigenetics, Histones and Modified Histones, Loading Controls
Primary or Secondary Primary
Gene ID (Entrez) 126961
Target Species Human
Content And Storage Store at -20C short term. Aliquot and store at -80C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.