Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Histone H3 Antibody, Novus Biologicals™
SDP

Catalog No. NB100747 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.125 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB100747 0.125 mg
NB017816 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB100747 Supplier Novus Biologicals Supplier No. NB100747
Add to Cart
Add to Cart

Sheep Polyclonal Antibody has been used in 6 publications

Histone H3 Polyclonal specifically detects Histone H3 in All species, Bovine, Human samples. It is validated for Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Single Cell Western, Western Blot.

Specifications

Antigen Histone H3
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Single Cell Western, Western Blot
Classification Polyclonal
Concentration 2.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:300, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence, Immunoprecipitation 1:1000, Immunohistochemistry-Paraffin 1:100, Single Cell Western 100 ug/mL
Formulation PBS with No Preservative
Gene Alias H3, H3 histone, family 3A, H3.3A, H3.3B, H3F2, H3F3, H3F3B, H3FM, HIST2H3C, histone H3.3, MGC87782, MGC87783
Gene Symbols H3C14
Host Species Sheep
Immunogen This Histone H3 antibody was raised against intact calf histone H3 at 15.4 kD, 136 amino acids, complexed with RNA.
Molecular Weight of Antigen 15 kDa
Purification Method Protein G purified
Quantity 0.125 mg
Regulatory Status RUO
Research Discipline Chromatin Modifiers, Chromatin Research, Epigenetics, Histones and Modified Histones, Loading Controls
Primary or Secondary Primary
Gene ID (Entrez) 126961
Test Specificity This Histone H3 antibody exhibits minimal cross reactivity with linker histone H1 and no cross-reactivity with histone core proteins H2A, H2B, or H4.
Target Species All species, Bovine, Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.