Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Histone H3 Antibody, Novus Biologicals™

Sheep Polyclonal Antibody has been used in 6 publications
$215.00 - $442.00
Specifications
Antigen | Histone H3 |
---|---|
Concentration | 2.5 mg/mL |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
Histone H3 Polyclonal specifically detects Histone H3 in Human, All Species, Bovine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Single Cell Western.Specifications
Histone H3 | |
Polyclonal | |
Purified | |
RUO | |
Bovine, Human, All species | |
H3, H3 histone, family 3A, H3.3A, H3.3B, H3F2, H3F3, H3F3B, H3FM, HIST2H3C, histone H3.3, MGC87782, MGC87783 | |
This Histone H3 antibody was raised against intact calf histone H3 at 15.4 kD, 136 amino acids, complexed with RNA. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. | |
15 kDa |
2.5 mg/mL | |
Unconjugated | |
Sheep | |
Chromatin Modifiers, Chromatin Research, Epigenetics, Histones and Modified Histones, Loading Controls | |
PBS with No Preservative | |
126961 | |
IgG | |
Protein G purified | |
This Histone H3 antibody exhibits minimal cross reactivity with linker histone H1 and no cross-reactivity with histone core proteins H2A, H2B, or H4. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title