Learn More
Description
Specifications
Specifications
| Antigen | Histone H3 (Dimethyl Lys36) |
| Applications | Western Blot, ELISA, Dot Blot, ChIP Assay |
| Classification | Monoclonal |
| Clone | C.15200182 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:1000 - 1:2000, ELISA 1:3000, Dot Blot 1:10000, Chromatin Immunoprecipitation (ChIP) 1-2 ug/IP |
| Formulation | PBS with 0.05% Sodium Azide |
| Gene Alias | H3, H3 histone, family 3A, H3.3A, H3.3B, H3F2, H3F3, H3F3B, H3FM, H3K36Me2, HIST2H3C, histone H3.3, MGC87782, MGC87783 |
| Gene Symbols | H3C14 |
| Host Species | Mouse |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
