Learn More
Description
Specifications
Specifications
| Antigen | Histone H3 (Dimethyl Lys4) |
| Applications | Western Blot, ELISA, Immunocytochemistry, Immunofluorescence, Dot Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:1000, ELISA 1:500, Immunocytochemistry/Immunofluorescence, Dot Blot 1:20000, Chromatin Immunoprecipitation (ChIP) 0.5-5 ug, Chromatin Immunoprecipitation Sequencing |
| Formulation | PBS with 0.05% Sodium Azide and 0.05% ProClin 300 |
| Gene Alias | H3, H3 histone, family 3A, H3.3A, H3.3B, H3F2, H3F3, H3F3B, H3FM, H3K4Me2, HIST2H3C, histone H3.3, MGC87782, MGC87783 |
| Gene Symbols | H3C14 |
| Host Species | Rabbit |
| Immunogen | The exact sequence of the immunogen to this Histone H3 [Dimethyl Lys4] antibody is proprietary. |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
