Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Histone H3, Monomethyl Lys4, p Thr6 Antibody, Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NB211037W Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB211037W 0.05 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB211037W Supplier Novus Biologicals Supplier No. NB211037AF405
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Histone H3 Polyclonal specifically detects Histone H3 in Human, Mouse, C. elegans samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Histone H3 (Monomethyl Lys4, p Thr6)
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 405
Dilution Western Blot, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Accession No. Q71DI3
Gene Alias H3, H3 histone, family 3A, H3.3A, H3.3B, H3F2, H3F3, H3F3B, H3FM, HIST2H3C, histone H3.3, MGC87782, MGC87783
Gene Symbols H3C14
Host Species Rabbit
Immunogen This Histone H3 [Monomethyl Lys4, p Thr6] antibody was raised against synthetic monomethylated/phosphorylated peptides surrounding Lysine 4 and Threonine 6 of human Histone H3.2 [Swiss Prot Q71DI3].
Purification Method Affinity Purified
Quantity 0.05 mL
Regulatory Status RUO
Research Discipline Chromatin Modifiers, Chromatin Research, Epigenetics, Histones and Modified Histones, Loading Controls
Primary or Secondary Primary
Gene ID (Entrez) 126961
Test Specificity Human, mouse, and S. cerevisiae. Predicted to react with many species including rat, chicken, Xenopus, Drosophila, and plant based on 100% sequence homology.
Target Species Human, Mouse, C. elegans
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.