Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Histone H3, p Ser10, ac Lys9 Antibody, Novus Biologicals™
SDP

Catalog No. NB310476 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.05 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB310476 0.025 mg
NB310477 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB310476 Supplier Novus Biologicals Supplier No. NB211095SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Histone H3 Polyclonal specifically detects Histone H3 in Human, Mouse samples. It is validated for Dot Blot, Immunocytochemistry/Immunofluorescence, Western Blot.

Specifications

Antigen Histone H3
Applications Dot Blot, Immunocytochemistry/Immunofluorescence, Western Blot
Classification Polyclonal
Concentration 0.25 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/mL, Immunocytochemistry/ Immunofluorescence 1:10, Dot Blot
Formulation PBS with 30% Glycerol
Gene Alias H3, H3 histone, family 3A, H3.3A, H3.3B, H3F2, H3F3, H3F3B, H3FM, HIST2H3C, histone H3.3, MGC87782, MGC87783
Host Species Rabbit
Immunogen Anti-Histone H3 [ac Lys9/phospho Ser10] affinity purified antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide with a acetylation surrounding Lysine 9 and a phosphorylation surrounding serine 10 of human Histone H3.
Purification Method Immunogen affinity purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Chromatin Modifiers, Chromatin Research, Epigenetics, Histones and Modified Histones, Loading Controls
Primary or Secondary Primary
Gene ID (Entrez) 126961
Target Species Human, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.