Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Histone H3, p Ser10, ac Lys9 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$213.73 - $346.83
Specifications
| Antigen | Histone H3 |
|---|---|
| Concentration | 0.25 mg/mL |
| Dilution | Western Blot 1.0 ug/mL, Immunocytochemistry/ Immunofluorescence 1:10, Dot Blot |
| Applications | Western Blot, Immunocytochemistry, Dot Blot |
| Classification | Polyclonal |
Description
Histone H3 Polyclonal antibody specifically detects Histone H3 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Dot BlotSpecifications
| Histone H3 | |
| Western Blot 1.0 ug/mL, Immunocytochemistry/ Immunofluorescence 1:10, Dot Blot | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| H3, H3 histone, family 3A, H3.3A, H3.3B, H3F2, H3F3, H3F3B, H3FM, HIST2H3C, histone H3.3, MGC87782, MGC87783 | |
| Anti-Histone H3 [ac Lys9/phospho Ser10] affinity purified antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide with a acetylation surrounding Lysine 9 and a phosphorylation surrounding serine 10 of human Histone H3. | |
| Immunogen affinity purified |
| 0.25 mg/mL | |
| Western Blot, Immunocytochemistry, Dot Blot | |
| Unconjugated | |
| Rabbit | |
| Chromatin Modifiers, Chromatin Research, Epigenetics, Histones and Modified Histones, Loading Controls | |
| PBS with 30% Glycerol | |
| 126961 | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title