Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Histone H3, p Ser10 Antibody (RM162), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB294385 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB294385 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB294385 Supplier Novus Biologicals Supplier No. NBP326022MFV450
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Histone H3 Monoclonal antibody specifically detects Histone H3 in Human, Vertebrate samples. It is validated for Western Blot, ELISA, Multiplex Immunoassay

Specifications

Antigen Histone H3
Applications Western Blot, ELISA, Multiplex
Classification Monoclonal
Clone RM162
Conjugate mFluor Violet 450 SE
Dilution Western Blot, ELISA, Multiplex Immunoassay
Formulation 50mM Sodium Borate
Gene Alias H3, H3 histone, family 3A, H3.3A, H3.3B, H3F2, H3F3, H3F3B, H3FM, HIST2H3C, histone H3.3, MGC87782, MGC87783
Host Species Rabbit
Immunogen This Histone H3 [p Ser10, Trimethyl Lys9] antibody (RM162) was raised against a trimethyl-phospho-peptide corresponding to Histone H3 [p Ser10, Trimethyl Lys9]
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Chromatin Modifiers, Chromatin Research, Epigenetics, Histones and Modified Histones, Loading Controls
Primary or Secondary Primary
Gene ID (Entrez) 126961
Target Species Human, Vertebrates
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.