Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Histone H3, p Ser28 Antibody (117C826), DyLight 405, Novus Biologicals™
SDP

Catalog No. NB10056595V Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056595V 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10056595V Supplier Novus Biologicals Supplier No. NB10056595V
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Histone H3 (p Ser28) Monoclonal specifically detects Histone H3 (p Ser28) in Fish, Human samples. It is validated for Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot.

Specifications

Antigen Histone H3 (p Ser28)
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 117C826
Conjugate DyLight 405
Dilution Western Blot, Immunocytochemistry/Immunofluorescence
Gene Accession No. Q71DI3
Gene Alias H3, H3 histone, family 3A, H3.3A, H3.3B, H3F2, H3F3, H3F3B, H3FM, HIST2H3C, histone H3.3, MGC87782, MGC87783
Gene Symbols H3C14
Host Species Mouse
Immunogen This Histone H3 [p Ser28] antibody (117C826) was raised against a synthetic peptide containing phospho serine at position 28 CARKS*APATGGVKK of human Histone H3. The immunogen sequence is conserved across mammalian and invertebrates, as well as plants.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Chromatin Modifiers, Chromatin Research, Epigenetics, Histones and Modified Histones, Loading Controls
Primary or Secondary Primary
Gene ID (Entrez) 126961
Test Specificity In anti-Fas mAb treated Jurkat, an 18 kDa band is observed using Histone H3 [p Ser28] antibody (117C826).
Target Species Fish, Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.