Learn More
Description
Specifications
Specifications
| Antigen | Histone H3 (p Thr6, Dimethyl Lys9) |
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence, Dot Blot, ChIP Assay |
| Classification | Polyclonal |
| Concentration | 0.49 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500, Immunocytochemistry/Immunofluorescence 1:100, Dot Blot 1:1000, Chromatin Immunoprecipitation (ChIP) 2-5 ug per million cells |
| Formulation | PBS and 30% Glycerol with 0.05% Sodium Azide |
| Gene Accession No. | Q71DI3, P84228 |
| Gene Alias | H3, H3 histone, family 3A, H3.3A, H3.3B, H3F2, H3F3, H3F3B, H3FM, H3K9Me2, H3T6p, HIST2H3C, histone H3.3, MGC87782, MGC87783 |
| Gene Symbols | H3C14 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
