Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Histone H3, p Thr6, Trimethyl Lys9 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB211054 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.05 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB211054 0.05 mg
NB211054SS 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB211054 Supplier Novus Biologicals Supplier No. NB211054
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Histone H3 Polyclonal specifically detects Histone H3 in Human, Mouse, C. elegans samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Immunocytochemistry/Immunofluorescence, Dot Blot.

Specifications

Antigen Histone H3 (p Thr6, Trimethyl Lys9)
Applications Western Blot, Immunocytochemistry, Immunofluorescence, Dot Blot, ChIP Assay
Classification Polyclonal
Concentration 0.55 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500, Immunocytochemistry/Immunofluorescence 1:200, Dot Blot 1:1000, Chromatin Immunoprecipitation (ChIP) 2-5 ug per million cells
Formulation PBS and 30% Glycerol with 0.05% Sodium Azide
Gene Accession No. Q71DI3, P84228
Gene Alias H3, H3 histone, family 3A, H3.3A, H3.3B, H3F2, H3F3, H3F3B, H3FM, H3T6p, HIST2H3C, histone H3.3, MGC87782, MGC87783
Gene Symbols H3C14
Host Species Rabbit
Immunogen This Histone H3 [p Thr6, Trimethyl Lys9] antibody was raised against synthetic trimethylated/phosphorylated peptide surrounding Lysine 9/ Threonine 6 of human Histone H3.2 [Swiss Prot Q71DI3].
Molecular Weight of Antigen 15 kDa
Purification Method Affinity Purified
Quantity 0.05 mg
Regulatory Status RUO
Research Discipline Chromatin Modifiers, Chromatin Research, Epigenetics, Histones and Modified Histones, Loading Controls
Primary or Secondary Primary
Gene ID (Entrez) 126961
Test Specificity Human, mouse, and S. cerevisiae. Predicted to react with many species including rat, chicken, Xenopus, Drosophila, and plant based on 100% sequence homology.
Target Species Human, Mouse, C. elegans
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.