Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Histone H3 Rabbit anti-Human, KHL, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
$182.31 - $457.74
Specifications
Antigen | Histone H3 |
---|---|
Applications | Dot Blot, Western Blot |
Classification | Polyclonal |
Conjugate | KLH |
Form | Purified |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
ABE288225MI
|
MilliporeSigma
ABE288225UL |
25 μL |
Each for $182.31
|
|
|||||
ABE288210MI
|
MilliporeSigma
ABE2882100UL |
100 μL |
Each for $457.74
|
|
|||||
Description
Histone H3 Polyclonal antibody specifically detects Histone H3 in Human samples. It is validated for Dot Blot, Western Blotting.Specifications
Histone H3 | |
Polyclonal | |
Purified | |
Epigenetics and Nuclear Function | |
NP_003520 | |
HIST1H3A, H3FA, HIST1H3B, HIST1H3C, HIST1H3D, HIST1H3E, HIST1H3F, HIST1H3G, HIST1H3H, HIST1H3I, HIST1H3J | |
IgG | |
Affinity Purified | |
This rabbit polyclonal antibody specifically detects Histone H3 with K14M substitution. |
Dot Blot, Western Blot | |
KLH | |
Rabbit | |
Human | |
Histone H3/a;Histone H3/b;Histone H3/c;Histone H3/d;Histone H3/f;Histone H3/h;Histone H3/I;Histone H3/j;Histone H3/k;Histone H3/l | |
KLH-conjugated linear peptide corresponding to 11 amino acids from the N-terminal region of Human Histone H3 with lysine 14 substituted with methionine. | |
Primary | |
Stable for 1 year at -20°C from date of receipt. | |
∼19 kDa observed; 15.40 kDa calculated. Uncharacterized bands may be observed in some lysate(s). |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title