Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Histone H3 Rabbit anti-Human, Mouse, Rat, Clone: 8V6Q6, Novus Biologicals™
SDP

Catalog No. NB166962 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB166962 20 μg
NB166961 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB166962 Supplier Novus Biologicals Supplier No. NBP31593320UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Histone H3 Monoclonal antibody specifically detects Histone H3 in Human, Mouse, Rat samples. It is validated for Western Blot, ChIP assay, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen Histone H3
Applications ChIP Assay, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 8V6Q6
Conjugate Unconjugated
Dilution Western Blot, Chromatin Immunoprecipitation, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias H3, H3 histone, family 3A, H3.3A, H3.3B, H3F2, H3F3, H3F3B, H3FM, HIST2H3C, histone H3.3, MGC87782, MGC87783
Host Species Rabbit
Immunogen A synthetic acetylated peptide around K27 of human Histone H3 (P68431).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Chromatin Modifiers, Chromatin Research, Epigenetics, Histones and Modified Histones, Loading Controls
Primary or Secondary Primary
Gene ID (Entrez) 126961
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.