Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Histone H3, Trimethyl Lys27 Antibody, Novus Biologicals™
SDP

Catalog No. NBP216840 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP216840 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP216840 Supplier Novus Biologicals Supplier No. NBP216840
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 2 publications

Histone H3 Polyclonal specifically detects Histone H3 in Human, Mouse, Rat, Zebrafish samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Dot Blot, Electron Microscopy, Immunohistochemistry Whole-Mount.

Specifications

Antigen Histone H3 (Trimethyl Lys27)
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.24mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000, Immunohistochemistry-Frozen 1:100-1:1000, Dot Blot 1:100 - 1:2000, Electron Microscopy 1:10 - 1:500, Immunohistochemistry Whole-Mount 1:100-1:500, Chromatin Immunoprecipitation (ChIP)
Formulation PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal
Gene Alias H3, H3 histone, family 3A, H3.3A, H3.3B, H3F2, H3F3, H3F3B, H3FM, H3K27Me3, H3pS28K27Me3, HIST2H3C, histone H3.3, MGC87782, MGC87783
Gene Symbols H3C14
Host Species Rabbit
Immunogen Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of human Histone H3K27me3 (Tri-methyl Lys27). The exact sequence is proprietary.
Molecular Weight of Antigen 15 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Chromatin Modifiers, Chromatin Research, Epigenetics, Histones and Modified Histones, Loading Controls
Primary or Secondary Primary
Gene ID (Entrez) 126961
Target Species Human, Mouse, Rat, Zebrafish
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.