Learn More
Description
Specifications
Specifications
| Antigen | Histone H3 (Trimethyl Lys37) |
| Applications | Western Blot, Dot Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Conjugate | DyLight 405 |
| Dilution | Western Blot 1ug/ml, Dot Blot 1:1000, ICC/IF (Negative) |
| Formulation | 50mM Sodium Borate with 0.05% Sodium Azide |
| Gene Alias | H3, H3 histone, family 3A, H3.3A, H3.3B, H3F2, H3F3, H3F3B, H3FM, HIST2H3C, histone H3.3, MGC87782, MGC87783 |
| Gene Symbols | H3C14 |
| Host Species | Rabbit |
| Immunogen | This Histone H3 [Trimethyl Lys37] antibody was raised against synthetic trimethylated peptide surrounding Lysine 37 of human Histone H3 [Swiss Prot# Q71DI3]. |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
