Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Histone H3, Trimethyl Lys9 Antibody (6F12-H4), Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NBP130141Y Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP130141Y 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP130141Y Supplier Novus Biologicals Supplier No. NBP130141AF647
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Histone H3 Monoclonal specifically detects Histone H3 in Human, Mouse, Rat, Porcine, C. elegans, Drosophila, Invertebrate, Mammal, Yeast samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen Histone H3 (Trimethyl Lys9)
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 6F12-H4
Conjugate Alexa Fluor 647
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Gene Accession No. P84243
Gene Alias H3, H3 histone, family 3A, H3.3A, H3.3B, H3F2, H3F3, H3F3B, H3FM, HIST2H3C, histone H3.3, MGC87782, MGC87783
Gene Symbols H3C14
Host Species Mouse
Immunogen This Histone H3 [Trimethyl Lys9] antibody (6F12-H4) was raised against a synthetic peptide made to an N-terminal region of Histone H3 (between amino acids 1-50). [UniProt# P84243]
Molecular Weight of Antigen 15 kDa
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Chromatin Modifiers, Chromatin Research, Epigenetics, Histones and Modified Histones, Loading Controls
Primary or Secondary Primary
Gene ID (Entrez) 126961
Test Specificity All mammals including human, mouse and rat, Drosophila, C. elegans, yeast. Invertebrate/Blattella germanica (German cockroach) reactivity reported in scientific literature (PMID: 23872316).
Target Species Human, Mouse, Rat, Pig, C. elegans, Drosophila, Invertebrate, Mammalia, Yeast
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.