Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Histone H4, ac Lys16 Antibody (C.15200219), Novus Biologicals™
SDP

Catalog No. NBP259242 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP259242 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP259242 Supplier Novus Biologicals Supplier No. NBP259242
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Histone H4 Monoclonal specifically detects Histone H4 in Human samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Dot Blot.

Specifications

Antigen Histone H4 (ac Lys16)
Applications Western Blot, Dot Blot, ChIP Assay
Classification Monoclonal
Clone C.15200219
Conjugate Unconjugated
Dilution Western Blot 1:500, Dot Blot 1:10000, Chromatin Immunoprecipitation (ChIP) 1-2 ug
Formulation PBS with 0.05% Sodium Azide
Gene Alias H4/A, H4/B, H4/C, H4/D, H4/E, H4/G, H4/H, H4/I, H4/J, H4/K, H4/M, H4/N, H4/O, H4/p, H4F2, H4FA, H4FB, H4FC, H4FD, H4FE, H4FG, H4FH, H4FI, H4FJ, H4FK, H4FM, H4FN, H4FO, H4K16ac, HIST1H4A, HIST1H4B, HIST1H4C, HIST1H4D, HIST1H4E, HIST1H4F, HIST1H4H, HIST1H4I, HIST1H4J, HIST1H4K, HIST1H4L, HIST2H4, HIST2H4A, HIST2H4B, histone 4, H4, histone cluster 4, H4, histone H4, MGC24116
Gene Symbols HIST4H4
Host Species Mouse
Immunogen The exact immunogen is propietary information.
Purification Method Protein A purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Chromatin Modifiers, Chromatin Research, DNA Repair, DNA replication Transcription Translation and Splicing, Epigenetics, HA Epitope Tags, Histones and Modified Histones
Primary or Secondary Primary
Gene ID (Entrez) 121504
Target Species Human
Content And Storage Store at -20C short term. Aliquot and store at -80C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.