Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Histone H4, Methyl Lys20 Antibody (C.15200147), Novus Biologicals™
SDP

Catalog No. NBP259236 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP259236 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP259236 Supplier Novus Biologicals Supplier No. NBP259236

Mouse Monoclonal Antibody

Histone H4 (Methyl Lys20) Monoclonal antibody specifically detects Histone H4 (Methyl Lys20) in Human, Mouse samples. It is validated for Western Blot, ChIP assay, Immunocytochemistry, Immunofluorescence, Dot Blot.

Specifications

Antigen Histone H4 (Methyl Lys20)
Applications Western Blot, Immunocytochemistry, Immunofluorescence, Dot Blot, ChIP Assay
Classification Monoclonal
Clone C.15200147
Conjugate Unconjugated
Dilution Western Blot 1:1000, Immunocytochemistry/Immunofluorescence, Dot Blot 1:20000, Chromatin Immunoprecipitation (ChIP) 0.5-1 ug/ChIP, Chromatin Immunoprecipitation Sequencing 0.5 - 1 ug/ChIP
Formulation PBS with 0.05% Sodium Azide
Gene Alias H4/A, H4/B, H4/C, H4/D, H4/E, H4/G, H4/H, H4/I, H4/J, H4/K, H4/M, H4/N, H4/O, H4/p, H4F2, H4FA, H4FB, H4FC, H4FD, H4FE, H4FG, H4FH, H4FI, H4FJ, H4FK, H4FM, H4FN, H4FO, H4K20Me, HIST1H4A, HIST1H4B, HIST1H4C, HIST1H4D, HIST1H4E, HIST1H4F, HIST1H4H, HIST1H4I, HIST1H4J, HIST1H4K, HIST1H4L, HIST2H4, HIST2H4A, HIST2H4B, histone 4, H4, histone cluster 4, H4, histone H4, MGC24116
Gene Symbols HIST4H4
Host Species Mouse
Immunogen The exact immunogen is propietary information.
Purification Method Protein A purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Chromatin Modifiers, Chromatin Research, DNA Repair, DNA replication Transcription Translation and Splicing, Epigenetics, HA Epitope Tags, Histones and Modified Histones
Primary or Secondary Primary
Gene ID (Entrez) 121504
Target Species Human, Mouse
Content And Storage Store at -20C short term. Aliquot and store at -80C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.