Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Histone H4, Monomethyl Arg3 Antibody, HRP, Novus Biologicals™
SDP

Catalog No. NB212011H Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB212011H 0.05 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB212011H Supplier Novus Biologicals Supplier No. NB212011H
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Histone H4 Polyclonal specifically detects Histone H4 in Human, Mouse, Rat, Mammal samples. It is validated for Western Blot, Dot Blot.

Specifications

Antigen Histone H4 (Monomethyl Arg3)
Applications Western Blot, Dot Blot
Classification Polyclonal
Conjugate HRP
Dilution Western Blot, Dot Blot
Formulation PBS with No Preservative
Gene Accession No. P62805
Gene Alias H4/A, H4/B, H4/C, H4/D, H4/E, H4/G, H4/H, H4/I, H4/J, H4/K, H4/M, H4/N, H4/O, H4/p, H4F2, H4FA, H4FB, H4FC, H4FD, H4FE, H4FG, H4FH, H4FI, H4FJ, H4FK, H4FM, H4FN, H4FO, HIST1H4A, HIST1H4B, HIST1H4C, HIST1H4D, HIST1H4E, HIST1H4F, HIST1H4H, HIST1H4I, HIST1H4J, HIST1H4K, HIST1H4L, HIST2H4, HIST2H4A, HIST2H4B, histone 4, H4, histone cluster 4, H4, histone H4, MGC24116
Gene Symbols HIST4H4
Host Species Rabbit
Immunogen Synthetic monomethylated peptide surrounding Arginine 3 of human Histone H4 [Swiss Prot P62805].
Purification Method Affinity Purified
Quantity 0.05 mL
Regulatory Status RUO
Research Discipline Chromatin Modifiers, Chromatin Research, DNA Repair, DNA replication Transcription Translation and Splicing, Epigenetics, HA Epitope Tags, Histones and Modified Histones
Primary or Secondary Primary
Gene ID (Entrez) 121504
Test Specificity Human, mouse and rat. Predicted to react with most mammal species.
Target Species Human, Mouse, Rat, Mammalia
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.