Learn More
Histone H4, Monomethyl Lys20 Antibody (5E10-D8), HRP, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | Histone H4 (Monomethyl Lys20) |
| Applications | Western Blot, ELISA, Dot Blot |
| Classification | Monoclonal |
| Clone | 5E10-D8 |
| Conjugate | HRP |
| Dilution | Western Blot, ELISA, Dot Blot |
| Gene Accession No. | P62805 |
| Gene Alias | H4/A, H4/B, H4/C, H4/D, H4/E, H4/G, H4/H, H4/I, H4/J, H4/K, H4/M, H4/N, H4/O, H4/p, H4F2, H4FA, H4FB, H4FC, H4FD, H4FE, H4FG, H4FH, H4FI, H4FJ, H4FK, H4FM, H4FN, H4FO, HIST1H4A, HIST1H4B, HIST1H4C, HIST1H4D, HIST1H4E, HIST1H4F, HIST1H4H, HIST1H4I, HIST1H4J, HIST1H4K, HIST1H4L, HIST2H4, HIST2H4A, HIST2H4B, histone 4, H4, histone cluster 4, H4, histone H4, MGC24116 |
| Gene Symbols | HIST4H4 |
| Host Species | Mouse |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.