Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Histone H4, Trimethyl Lys20 Antibody (RM208), DyLight 488, Novus Biologicals™
SDP

Catalog No. NB294336 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB294336 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB294336 Supplier Novus Biologicals Supplier No. NBP326024G
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Histone H4 Monoclonal antibody specifically detects Histone H4 in Human, Vertebrate samples. It is validated for Western Blot, ELISA, Multiplex Immunoassay

Specifications

Antigen Histone H4
Applications Western Blot, ELISA, Multiplex
Classification Monoclonal
Clone RM208
Conjugate DyLight 488
Dilution Western Blot, ELISA, Multiplex Immunoassay
Formulation 50mM Sodium Borate
Gene Alias H4/A, H4/B, H4/C, H4/D, H4/E, H4/G, H4/H, H4/I, H4/J, H4/K, H4/M, H4/N, H4/O, H4/p, H4F2, H4FA, H4FB, H4FC, H4FD, H4FE, H4FG, H4FH, H4FI, H4FJ, H4FK, H4FM, H4FN, H4FO, HIST1H4A, HIST1H4B, HIST1H4C, HIST1H4D, HIST1H4E, HIST1H4F, HIST1H4H, HIST1H4I, HIST1H4J, HIST1H4K, HIST1H4L, HIST2H4, HIST2H4A, HIST2H4B, histone 4, H4, histone cluster 4, H4, histone H4, MGC24116
Host Species Rabbit
Immunogen A trimethyl-peptide corresponding to Histone H4 [Methyl Lys20]
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Chromatin Research, DNA Repair, DNA replication Transcription Translation and Splicing, Epigenetics, HA Epitope Tags, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 121504
Target Species Human, Vertebrates
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.