Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Histone H4K16ac Polyclonal Antibody, Active Motif™

Rabbit Polyclonal Antibody
$159.90 - $651.90
Specifications
| Antigen | Histone H4K16ac |
|---|---|
| Concentration | Conc. Not Determined |
| Applications | ChIP Assay, ChIP sequencing (ChIP-seq), Dot Blot, ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-199-3936
![]()
|
Active Motif
39168 |
10 μL |
Each for $159.90
|
|
|||||
50-199-3934
![]()
|
Active Motif
39068 |
50 μL |
Each for $375.15
|
|
|||||
50-199-3935
![]()
|
Active Motif
39167 |
100 μL |
Each for $651.90
|
|
|||||
Description
Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes. Two molecules of each of the four core histones (H2A, H2B, H3, and H4) form an octamer, around which approximately 146 bp of DNA is wrapped in repeating units, called nucleosomes. The linker histone, H1, interacts with linker DNA between nucleosomes and functions in the compaction of chromatin into higher order structures. This gene is intronless and encodes a member of the histone H4 family. Transcripts from this gene lack polyA tails but instead contain a palindromic termination element. This gene is found in the large histone gene cluster on chromosome 6.Specifications
| Histone H4K16ac | |
| ChIP Assay, ChIP sequencing (ChIP-seq), Dot Blot, ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Drosophila, Human, Mouse, Yeast | |
| P62805, P62806 | |
| 121504, 319158, 326619, 326620, 41773, 8294, 8359, 8366, 8367, 8370, 97122 | |
| This Histone H4 acetyl Lys16 antibody was raised against a peptide including acetyl-lysine 16 of histone H4. | |
| Primary | |
| Antibody |
| Conc. Not Determined | |
| Polyclonal | |
| Liquid | |
| RUO | |
| whole serum with 30% glycerol and 0.035% sodium azide; pH | |
| acetyl Histone H4; Acetyl-H4 K16; Acetyl-H4 Lys16; Acetyl-Histone H4 K16; Acetyl-Histone H4 Lys16; FO108; H4; H4 histone family, member A; H4 histone family, member I; H4 histone family, member J; H4 histone family, member M; H4 histone family, member N; H4 histone, family 2; H4 K16ac; H4/A; H4/B; H4/C; H4/D; H4/E; H4/G; H4/H; H4/I; H4/J; H4/K; H4/M; H4/N; H4/O; H4/p; H4-12; H4-16; H4-53; H4C1; H4C11; H4C12; H4C13; H4C14; H4C15; H4C2; H4c3; H4c4; H4C5; H4C6; H4C8; H4C9; H4f16; H4F2; H4FA; H4FB; H4FC; H4FD; H4FE; H4FG; H4FH; H4FI; H4FJ; H4FK; H4FM; H4FN; H4FO; H4K16; H4K16a; H4Lys16ac; H4M; HIST1H4; HIST1H4A; Hist1h4b; Hist1h4c; HIST1H4D; HIST1H4E; HIST1H4F; HIST1H4H; HIST1H4I; HIST1H4J; HIST1H4K; HIST1H4L; Hist1h4m; Hist2h4; Hist2h4a; HIST2H4B; HIST4H4; histone 1, H4a; histone 1, H4b; histone 1, H4e; histone 1, H4i; histone 2, H4; histone 2, H4a; Histone 4; Histone 4 family, member M; histone 4, H4; histone cluster 1, H4a; histone cluster 1, H4b; histone cluster 1, H4e; histone cluster 1, H4i; histone cluster 2, H4; histone cluster 2, H4a; Histone Cluster 4; histone cluster 4, H4; histone family member; Histone H4; Histone H4 acetyl; Histone H4A; Histone H4K16; Histone H4K16ac; histone IV, family 2; X04652 | |
| H4-16, H4C1, H4C14, H4C2, H4C5, H4C9, His4r | |
| IgG | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| H4C1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title