Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Histone H4K5ac/K8ac Recombinant Mouse Monoclonal Antibody, AbFlex™, Active Motif™

Mouse Recombinant Monoclonal Antibody
$108.45 - $482.00
Specifications
Antigen | Histone H4K5ac/K8ac |
---|---|
Concentration | 1 mg/mL |
Applications | ChIP sequencing (ChIP-seq), Dot Blot, Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-199-3819
![]() ![]() |
Active Motif
91320 |
10 μg |
Each for $108.45
|
|
|||||
50-199-3818
![]() ![]() |
Active Motif
91319 |
100 μg |
Each for $482.00
|
|
|||||
Description
Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes. Two molecules of each of the four core histones (H2A, H2B, H3, and H4) form an octamer, around which approximately 146 bp of DNA is wrapped in repeating units, called nucleosomes. The linker histone, H1, interacts with linker DNA between nucleosomes and functions in the compaction of chromatin into higher order structures. This gene is intronless and encodes a member of the histone H4 family. Transcripts from this gene lack polyA tails but instead contain a palindromic termination element. This gene is found in the large histone gene cluster on chromosome 6.Specifications
Histone H4K5ac/K8ac | |
ChIP sequencing (ChIP-seq), Dot Blot, Western Blot | |
Unconjugated | |
Mouse | |
Human, Multi-species, Mouse, Rat | |
P62804 | |
HIST1H4A, HIST1H4B, HIST2H4A | |
IgG1 | |
Protein A | |
Antibody |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
140mM HEPES, 32mM sodium acetate with 30% glycerol, 70mM NaCl and 0.035% sodium azide; pH 7.5 | |
502913, 64627, 679983 | |
This antibody was raised against a full-length human H4 protein containing acetyl k5 and k8 (H4k5ack8ac). | |
Primary | |
-20°C, Avoid Freeze/Thaw Cycles | |
HIST1H4B |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title