Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HistoneH2AE Rabbit anti-Human, Clone: SR4-17, Novus Biologicals™
SDP

Catalog No. NBP267337 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP267337 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP267337 Supplier Novus Biologicals Supplier No. NBP267337

Rabbit Monoclonal Antibody

Histone H2AE Monoclonal specifically detects Histone H2AE in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen HistoneH2AE
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone SR4-17
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50-1:200
Formulation TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide
Gene Alias H2A histone family, member M, H2A/m, H2AFA, H2AFMHistone H2A/m, HIST1H2AE, histone 1, H2ab, histone cluster 1, H2ab, histone H2A type 1-B/E, Histone H2A.2, Histone H2A/a
Gene Symbols HIST1H2AE
Host Species Rabbit
Immunogen Recombinant protein within Human Histone H2AE aa 1-83/ 130 (hydroxyl Y39) conjugated to Keyhole Limpet Haemocyanin (KLH). (SwissProt: P04908 Human; SwissProt: P28001 Human; SwissProt: Q93077 Human; SwissProt: P35065 Human; SwissProt: Q99878 Human; SwissProt: C0HKE6 Mouse; SwissProt: C0HKE1 Mouse; SwissProt: P02262 Rat)
Purification Method Protein A purified
Quantity 100 μL
Research Discipline Epigenetics, Histones and Modified Histones
Primary or Secondary Primary
Gene ID (Entrez) 3012
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.