Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HistoneH2B Rabbit anti-Human, Clone: SD20-63, Novus Biologicals™
SDP

Catalog No. NBP267753 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP267753 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP267753 Supplier Novus Biologicals Supplier No. NBP267753
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Histone H2B Monoclonal specifically detects Histone H2B in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.

Specifications

Antigen HistoneH2B
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
Classification Monoclonal
Clone SD20-63
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, Flow Cytometry 1:50-1:100, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:100-1:500, Immunoprecipitation, Immunohistochemistry-Paraffin 1:50-1:5000
Formulation TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide
Gene Alias FLJ35099, FLJ56780, FLJ56787, histone 2, H2bf, histone cluster 2, H2bf, histone H2B type 2-F, MGC131639
Gene Symbols HIST2H2BF
Host Species Rabbit
Immunogen Synthetic peptide within human Histone H2B aa 110 to the C-terminus (acetyl K121). (SwissProt: O60814 Human; SwissProt: P10854 Mouse; SwissProt: Q64475 Mouse; SwissProt: Q00729 Rat)
Purification Method Protein A purified
Quantity 100 μL
Research Discipline Chromatin Research, Epigenetics, Histones and Modified Histones
Primary or Secondary Primary
Gene ID (Entrez) 440689
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.