Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Histone H3 (Monomethyl Lys18) Rabbit anti-Human, Mouse, Rat, Clone: SA42-07, Novus Biologicals™
SDP

Catalog No. NBP267289 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP267289 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP267289 Supplier Novus Biologicals Supplier No. NBP267289

Rabbit Monoclonal Antibody

Histone H3 Monoclonal specifically detects Histone H3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen Histone H3 (Monomethyl Lys18)
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone SA42-07
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:5000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunohistochemistry-Paraffin 1:50-1:200
Formulation TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide
Gene Alias H3, H3 histone, family 3A, H3.3A, H3.3B, H3F2, H3F3, H3F3B, H3FM, HIST2H3C, histone H3.3, MGC87782, MGC87783
Gene Symbols H3C14
Host Species Rabbit
Immunogen Synthetic peptide within Human Histone H3 aa 1-50/ 136 (mono methyl K18) conjugated to Keyhole Limpet Haemocyanin (KLH). (SwissProt: P68431 Human; SwissProt: P84243 Human; SwissProt: Q71DI3 Human; SwissProt: Q16695 Human; SwissProt: Q6NXT2 Human; SwissProt: P68433 Mouse; SwissProt: Q6LED0 Rat)
Molecular Weight of Antigen 15 kDa
Purification Method Protein A purified
Quantity 100 μL
Research Discipline Chromatin Modifiers, Chromatin Research, Epigenetics, Histones and Modified Histones, Loading Controls
Primary or Secondary Primary
Gene ID (Entrez) 8290
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.