Learn More
Description
Cobalt-based IMAC medium offering different selectivity compared to nickel-charged media. Allows protein purification under native or denaturing conditions and can be used with prokaryotic and eukaryotic expression systems.
- Prepacked with cobalt-based chromatography media
- Binding capacity approximately 7mg MBP
- Maximum pressure: 3 bar
- Column material: polypropylene
- Compatible with commonly used buffers and easily regenerated with 0.5 M NaOH
- 1 and 5mL columns
Media
- Ligand: TALON™ Superflow Cobalt-IMAC
- Matrix: 6% cross-linked agarose
- Average particle size: 60 to 160μm
- pH stability working range: 3 to 12
1mL columns
- Column I.D.: 7mm
- Flow velocity: 1 to 4mL/min.
5mL columns
- Column I.D.: 16
- Flow velocity: 5 to 20mL/min.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Quantity | 1 x 1 mL |
| For Use With (Application) | For PurifiCatIon of Histidine-Tagged Recombinant Proteins by Immobilized Metal Affinity Chromatography, Used Buffers and Easily Regenerated with 0.5 M NaOH |
| Storage Requirements | 4°C to 8°C |
| Binding Capacity | Up to 20 mg Histidine-tagged Protein1 |
| Diameter (English) Inner | 0.27 in. |
| Product Type | HiTrap Talon crude prepacked Column |
| Diameter (Metric) Inner | 0.7 cm |
| Flow Rate | 1 to 4 mL/min |
| Particle Size | 60 to 160 μm |
| Material | Polypropylene |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
