Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HIV-1 Gag p24 Antibody (8G9), Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NB201050 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB201050 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB201050 Supplier Novus Biologicals Supplier No. NBP241336AF647
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HIV-1 Gag p24 Monoclonal antibody specifically detects HIV-1 Gag p24 in Virus samples. It is validated for Western Blot, ELISA

Specifications

Antigen HIV-1 Gag p24
Applications Western Blot, ELISA
Classification Monoclonal
Clone 8G9
Conjugate Alexa Fluor 647
Formulation 50mM Sodium Borate
Host Species Mouse
Immunogen Antibody was raised against a recombinant full-length HIV-1 Gag p24 protein. Amino Acid Squence: pivqniqgqmvhqaisprtlnawvkvveekafspevipmfsalsegatpqdlntmlntvgghqaamqmlketineeaaewdrvhpvhagpiapgqmreprgsdiagttstlqeqigwmtnnppipvgeiykrwiilglnkivrmysptsildirqgpkepfrdyvdrfyktlraeqasqevknwmtetllvqnanpdcktilkalgpaatleemmtacqgvggpghkarvla
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Infections (Virus Bacteria and Parasites)
Primary or Secondary Primary
Gene ID (Entrez) 155030
Target Species Virus
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.