Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HIV-1 Gag p24 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB017726 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
0.5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB017726 0.1 mL
NB10065514 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB017726 Supplier Novus Biologicals Supplier No. NB100655140.1ML
Add to Cart
Add to Cart

Goat Polyclonal Antibody

HIV-1 Gag p24 Polyclonal specifically detects HIV-1 Gag p24 in Virus samples. It is validated for Western Blot.

Specifications

Antigen HIV-1 p24
Applications Western Blot
Classification Polyclonal
Concentration 4.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:2000
Formulation Phosphate buffered saline with 0.01% Sodium Azide
Gene Alias Capsid protein p24, Human immunodeficiency virus 1, Human immunodeficiency virus type 1 p24
Host Species Goat
Immunogen Purified native p24, from strain IIIB.
Purification Method IgG purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 155348
Test Specificity Recognizes the HIV1 viral capsid p24 protein encoded by the gag gene along with p6, p7 and p17 which provide structural elements of the virus.
Target Species Virus
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.