Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HIV-1 Gag p24 Antibody (HIV1-24/661), Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NBP234636Y Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP234636Y 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP234636Y Supplier Novus Biologicals Supplier No. NBP234636AF647
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HIV-1 Gag p24 Monoclonal antibody specifically detects HIV-1 Gag p24 in Virus samples. It is validated for ELISA, Immunocytochemistry, Immunofluorescence.

Specifications

Antigen HIV-1 Gag p24
Applications ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone HIV1-24/661
Conjugate Alexa Fluor 647
Dilution ELISA, Immunocytochemistry/Immunofluorescence
Gene Accession No. P04585, P04585
Gene Alias Capsid protein p24, Human immunodeficiency virus 1, Human immunodeficiency virus type 1 p24
Gene Symbols gag
Host Species Mouse
Immunogen Recombinant HIV-1 Gag p24 protein (Uniprot: P04585)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 155030
Test Specificity Human immunodeficiency virus (HIV) is a retrovirus that causes acquired immune deficiency syndrome (AIDS), a condition in humans in which the immune system begins to fail, leading to life-threatening opportunistic infections. HIV mainly infects vital cells in the human immune system such as helper T cells (specifically CD4+ T cells), macrophages and dendritic cells. Two species of HIV infect humans: HIV-1 and HIV-2, with HIV-1 being the more virulent strain. The gag gene of human immunodeficiency virus 1 (HIV-1) encodes a precursor protein known as Pr55Gag. The viral protease PR cleaves this precursor to generate p17, p24, p7, and p6 proteins, which are required for virus particle assembly. HIV-1 Gag p24 is a capsid protein that constitutes the core of AIDS virus HIV-1. p6 and p7 are the components of the nucleocapsid, and p17 provides a protective matrix. HIV-1 Gag p24 is indispensable to the reproduction of AIDS virus and constitutes an essential element for the AIDS virus particle construction. As this protein is detectable from the early stage of AIDS virus infection, its measurement is commonly used as an indicator of HIV-1 infection and viral load.
Target Species Virus
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.