Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HIV-1 Gag p24 Antibody (ND1), Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NB500473Y Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB500473Y 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB500473Y Supplier Novus Biologicals Supplier No. NB500473AF647
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HIV-1 Gag p24 Monoclonal antibody specifically detects HIV-1 Gag p24 in Virus samples. It is validated for Western Blot, ELISA, Immunocytochemistry, Immunofluorescence.

Specifications

Antigen HIV-1 Gag p24
Applications Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone ND1
Concentration 1.0mg/mL
Conjugate Alexa Fluor 647
Dilution Western Blot, ELISA, Immunocytochemistry/Immunofluorescence
Gene Alias Capsid protein p24, Human immunodeficiency virus 1, Human immunodeficiency virus type 1 p24
Gene Symbols gag
Host Species Mouse
Immunogen HIV-1 Gag p24 C-terminal peptide.
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 155030
Test Specificity The antibody reacts with HIV-1 Gag p24). Specificity was verified by Western Blotting in reducing conditions with Sanofi Pasteur NEW LAV-BLOT I kit. Epitope was reported to be located within the amino acids 51-110.
Target Species Virus
Content And Storage Store at 4C in the dark.
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.